accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78751-80-1

IUPAC Name: 2-bromo-7-tert-butylpyrene
Synonyms: 2-bromo-7-tert-butylpyrene
Molecular Weight (g/mol): 337.26
Molecular Formula: C20H17Br
InChi Key: RYLYZVOQPGVCQT-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC2=CC=C3C=C(Br)C=C4C=CC(=C1)C2=C34
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results