accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 787618-22-8

IUPAC Name: [2',6'-bis(propan-2-yloxy)-[1,1'-biphenyl]-2-yl]dicyclohexylphosphane
Synonyms: dicyclohexyl({2',6'-diisopropoxy-[1,1'-biphenyl]-2-yl})phosphane
Molecular Weight (g/mol): 466.65
Molecular Formula: C30H43O2P
InChi Key: MXFYYFVVIIWKFE-UHFFFAOYSA-N
SMILES: CC(C)OC1=CC=CC(OC(C)C)=C1C1=CC=CC=C1P(C1CCCCC1)C1CCCCC1
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results