CAS RN 78851-84-0
IUPAC Name:
methyl 2-amino-5-[(diaminomethylidene)amino]pentanoate dihydrochloride
Synonyms:
methyl 2-amino-5-[(diaminomethylidene)amino]pentanoate dihydrochloride
Molecular Weight (g/mol):
261.15
Molecular Formula:
C7H18Cl2N4O2
InChi Key:
XQYZOBNLCUAXLF-UHFFFAOYNA-N
SMILES:
Cl.Cl.COC(=O)C(N)CCCN=C(N)N
1
–
1
of
1
results
1
D-Arginine Methyl Ester Dihydrochloride 98.0+%, TCI America™
CAS: 78851-84-0 Molecular Formula: C7H18Cl2N4O2 Molecular Weight (g/mol): 261.147 MDL Number: MFCD00153417 InChI Key: XQYZOBNLCUAXLF-ZJIMSODOSA-N PubChem CID: 12714066 IUPAC Name: methyl (2R)-2-amino-5-(diaminomethylideneamino)pentanoate;dihydrochloride SMILES: COC(=O)C(CCCN=C(N)N)N.Cl.Cl