accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNNOOH

CAS RN 78875-63-5

IUPAC Name: 4-phenyl-1,2,3-thiadiazole-5-carboxylic acid
Synonyms: 4-phenyl-1,2,3-thiadiazole-5-carboxylic acid
Molecular Weight (g/mol): 206.22
Molecular Formula: C9H6N2O2S
InChi Key: DQZLGXDVESJWKA-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(N=NS1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results