accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3COONHH2C

CAS RN 78888-18-3

IUPAC Name: tert-butyl N-(prop-2-en-1-yl)carbamate
Synonyms: tert-butyl N-(prop-2-en-1-yl)carbamate
Molecular Weight (g/mol): 157.21
Molecular Formula: C8H15NO2
InChi Key: AWARHXCROCWEAK-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC=C
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)