accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78902-09-7

IUPAC Name: 2-(2,2-diethoxyethyl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-(2,2-diethoxyethyl)isoindole-1,3-dione
Molecular Weight (g/mol): 263.29
Molecular Formula: C14H17NO4
InChi Key: GEFXJJJQUSEHLV-UHFFFAOYSA-N
SMILES: CCOC(CN1C(=O)C2=CC=CC=C2C1=O)OCC
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results