accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78950-36-4

IUPAC Name: 3-aminobenzene-1-carbothioamide
Synonyms: 3-aminobenzenecarbothioamide
Molecular Weight (g/mol): 152.22
Molecular Formula: C7H8N2S
InChi Key: ZKWTUTBIHCNCKU-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC(N)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results