accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHCH3O

CAS RN 79-16-3

IUPAC Name: N-methylacetamide
Synonyms: N-methylacetamide
Molecular Weight (g/mol): 73.1
Molecular Formula: C3H7NO
InChi Key: OHLUUHNLEMFGTQ-UHFFFAOYSA-N
SMILES: CNC(C)=O
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)