accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClO

CAS RN 79-36-7

IUPAC Name: 2,2-dichloroacetyl chloride
Synonyms: dichloroacetyl chloride chlorure de dichloracetyle chlorid kyseliny dichloroctove C2hcl3O
Molecular Weight (g/mol): 147.38
Molecular Formula: C2HCl3O
InChi Key: FBCCMZVIWNDFMO-UHFFFAOYSA-N
SMILES: ClC(Cl)C(Cl)=O
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)