accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH2NH2O

CAS RN 79-39-0

IUPAC Name: 2-methylprop-2-enamide
Synonyms: methacrylamide methacrylic amide α-methyl acrylic amide
Molecular Weight (g/mol): 85.11
Molecular Formula: C4H7NO
InChi Key: FQPSGWSUVKBHSU-UHFFFAOYSA-N
SMILES: CC(=C)C(N)=O
Molecular Weight (g/mol) 
  • (2)
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
Boiling Point 
  • (4)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (2)
  • (4)
  • (3)
  • (2)
  • (7)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)
  • (3)
  • (2)
  • (3)
  • (3)