accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 79-42-5

IUPAC Name: 2-sulfanylpropanoic acid
Synonyms: thiolactic acid|(S)-thiolactic acid|(s)-thiolactic acid|2-thiolpropionic acid|(R)-thiolactic acid|(r)-thiolactic acid|thiolactic acid, (R)-|thiolactic acid, (S)-|thiolactic acid, (r)-|thiolactic acid, (s)-
Molecular Weight (g/mol): 106.14
Molecular Formula: C3H6O2S
InChi Key: PMNLUUOXGOOLSP-UHFFFAOYNA-N
SMILES: CC(S)C(O)=O
Quantity 
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (4)
Boiling Point 
  • (2)
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (5)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (3)
  • (6)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (5)
  • (3)
  • (7)