accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NOO

CAS RN 79-46-9

IUPAC Name: 2-nitropropane
Synonyms: β-nitropropane
Molecular Weight (g/mol): 89.09
Molecular Formula: C3H7NO2
InChi Key: FGLBSLMDCBOPQK-UHFFFAOYSA-N
SMILES: CC(C)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)