accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 79-74-3

IUPAC Name: 2,5-bis(2-methylbutan-2-yl)benzene-1,4-diol
Synonyms: diamylhydroquinone
Molecular Weight (g/mol): 250.38
Molecular Formula: C16H26O2
InChi Key: CZNRFEXEPBITDS-UHFFFAOYSA-N
SMILES: CCC(C)(C)C1=CC(O)=C(C=C1O)C(C)(C)CC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results