accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCH3H3CCH3(E)

CAS RN 79-77-6

IUPAC Name: (3E)-4-(2,6,6-trimethylcyclohex-1-en-1-yl)but-3-en-2-one
Synonyms: β-ionone β-ionone (trans)
Molecular Weight (g/mol): 192.3
Molecular Formula: C13H20O
InChi Key: PSQYTAPXSHCGMF-BQYQJAHWSA-N
SMILES: CC(=O)\C=C\C1=C(C)CCCC1(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results