CAS RN 79029-81-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Supelco™ Chromosorb™ LC-9
Prepared by chemically bonding an amino type functional group on the microparticle silica substrate.