accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 79047-41-9

IUPAC Name: (2-butyl-4-chloro-1H-imidazol-5-yl)methanol
Synonyms: (2-butyl-5-chloro-3H-imidazol-4-yl)methanol
Molecular Weight (g/mol): 188.66
Molecular Formula: C8H13ClN2O
InChi Key: DXSZKDOOHOBZMT-UHFFFAOYSA-N
SMILES: CCCCC1=NC(Cl)=C(CO)N1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results