CAS RN 79060-88-1
Selectophore™ Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, ≥98.0% (based on Na, AAS), MilliporeSigma™ Supelco™
MDL Number: MFCD00043323 Synonym: NaBARF; Tetrakis[3,5-bis(trifluoromethyl)phenyl]boron sodium
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, 97%, may cont. 1-5% water
CAS: 79060-88-1 Molecular Formula: C32H12BF24Na Molecular Weight (g/mol): 886.209 MDL Number: MFCD00043323 InChI Key: LTGMONZOZHXAHO-UHFFFAOYSA-N Synonym: sodium tetrakis 3,5-bis trifluoromethyl phenyl borate,nabarf,sodium tetrakis 3,5-bis trifluoromethyl phenyl boranuide,sodium2-sulfonatoethylmethacrylate,sodium tetrakis 3,5-bis trifluoromethyl phenyl bor,sodium tetrakis 3,5-bis-trifluoromethylphenyl borate,sodiotetrakis 3,5-bis trifluoromethyl phenyl boron v,sodium tetrakis 3,5-bis trifluoromethyl phenyl-borate,sodium tetrakis 3,5-bis trifluoromethyl phenyl borate 1- PubChem CID: 23681909 IUPAC Name: sodium;tetrakis[3,5-bis(trifluoromethyl)phenyl]boranuide SMILES: [B-](C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)(C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)(C3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F)C4=CC(=CC(=C4)C(F)(F)F)C(F)(F)F.[Na+]
Sodium Tetrakis[3,5-bis(trifluoromethyl)phenyl]borate Hydrate 98.0+%, TCI America™
CAS: 79060-88-1 Molecular Formula: C32H12BF24Na Molecular Weight (g/mol): 886.209 MDL Number: MFCD00043323 InChI Key: LTGMONZOZHXAHO-UHFFFAOYSA-N Synonym: sodium tetrakis 3,5-bis trifluoromethyl phenyl borate,nabarf,sodium tetrakis 3,5-bis trifluoromethyl phenyl boranuide,sodium2-sulfonatoethylmethacrylate,sodium tetrakis 3,5-bis trifluoromethyl phenyl bor,sodium tetrakis 3,5-bis-trifluoromethylphenyl borate,sodiotetrakis 3,5-bis trifluoromethyl phenyl boron v,sodium tetrakis 3,5-bis trifluoromethyl phenyl-borate,sodium tetrakis 3,5-bis trifluoromethyl phenyl borate 1- PubChem CID: 23681909 IUPAC Name: sodium;tetrakis[3,5-bis(trifluoromethyl)phenyl]boranuide SMILES: [B-](C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)(C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)(C3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F)C4=CC(=CC(=C4)C(F)(F)F)C(F)(F)F.[Na+]
Sigma Aldrich Sodium Tetrakis[3,5-Bis(Trifluoromethyl)Phenyl]Borate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.