accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)OHHNCH3OOCH3CH3CH3

CAS RN 79069-14-0

IUPAC Name: tert-butyl N-[(2S)-1-hydroxy-3-methylbutan-2-yl]carbamate
Synonyms: tert-butyl N-[(2S)-1-hydroxy-3-methylbutan-2-yl]carbamate|N-BOC-L-VALINOL
Molecular Weight (g/mol): 203.28
Molecular Formula: C10H21NO3
InChi Key: OOQRRYDVICNJGC-MRVPVSSYSA-N
SMILES: CC(C)[C@@H](CO)NC(=O)OC(C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)