accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH3CFFF

CAS RN 79080-31-2

IUPAC Name: 1,5-dimethyl-3-(trifluoromethyl)-1H-pyrazole
Synonyms: 1,5-dimethyl-3-(trifluoromethyl)pyrazole
Molecular Weight (g/mol): 164.13
Molecular Formula: C6H7F3N2
InChi Key: LGTRXZPAYGKNLP-UHFFFAOYSA-N
SMILES: CN1N=C(C=C1C)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results