accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 79098-80-9

IUPAC Name: 1-(piperidin-4-yl)-1H-1,2,3-benzotriazole hydrochloride
Synonyms: 1-(piperidin-4-yl)-1,2,3-benzotriazole hydrochloride
Molecular Weight (g/mol): 238.72
Molecular Formula: C11H15ClN4
InChi Key: SIZIQJKLJZWLLD-UHFFFAOYSA-N
SMILES: Cl.C1CC(CCN1)N1N=NC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results