accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrS

CAS RN 791078-04-1

IUPAC Name: 2-[2-(bromomethyl)phenyl]thiophene
Synonyms: 2-[2-(bromomethyl)phenyl]thiophene
Molecular Weight (g/mol): 253.16
Molecular Formula: C11H9BrS
InChi Key: IJNVNLWIQXMBPA-UHFFFAOYSA-N
SMILES: BrCC1=CC=CC=C1C1=CC=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results