accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 79109-69-6

IUPAC Name: trimethyl(2-{5-[2-(trimethylsilyl)ethynyl]thiophen-2-yl}ethynyl)silane
Synonyms: trimethyl(2-{5-[2-(trimethylsilyl)ethynyl]thiophen-2-yl}ethynyl)silane
Molecular Weight (g/mol): 276.54
Molecular Formula: C14H20SSi2
InChi Key: RAPGDWBCCYLGCE-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#CC1=CC=C(S1)C#C[Si](C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results