accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 79151-78-3

NNNNNNFPFFFFFFPFFFFFCo

CAS RN 79151-78-3

IUPAC Name: λ²-cobalt(2+) tris(2,2'-bipyridine) bis(hexafluoro-λ⁵-phosphanuide)
Synonyms: λ²-cobalt(2+) tris(bipyridyl) dihexafluorophosphate
Molecular Weight (g/mol): 817.43
Molecular Formula: C30H24CoF12N6P2
InChi Key: IDXASOGRZNYSPC-UHFFFAOYSA-N
SMILES: [Co++].F[P-](F)(F)(F)(F)F.F[P-](F)(F)(F)(F)F.C1=CC=C(N=C1)C1=CC=CC=N1.C1=CC=C(N=C1)C1=CC=CC=N1.C1=CC=C(N=C1)C1=CC=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)