accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClMgBr

CAS RN 79175-35-2

IUPAC Name: bromo(3,4-dichlorophenyl)magnesium
Synonyms: bromo(3,4-dichlorophenyl)magnesium
Molecular Weight (g/mol): 250.2
Molecular Formula: C6H3BrCl2Mg
InChi Key: IRROSODEQHIZEV-UHFFFAOYSA-M
SMILES: ClC1=CC=C([Mg]Br)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results