CAS RN 79218-15-8
IUPAC Name:
tert-butyl but-2-enoate
Synonyms:
tert-butyl but-2-enoate
Molecular Weight (g/mol):
142.2
Molecular Formula:
C8H14O2
InChi Key:
QHSPZGZEUDEIQM-UHFFFAOYSA-N
SMILES:
CC=CC(=O)OC(C)(C)C
1
–
1
of
1
results
1
tert-Butyl Crotonate 98.0+%, TCI America™
CAS: 79218-15-8 Molecular Formula: C8H14O2 Molecular Weight (g/mol): 142.198 MDL Number: MFCD00059055 InChI Key: QHSPZGZEUDEIQM-AATRIKPKSA-N Synonym: Crotonic Acid tert-Butyl Ester PubChem CID: 5463127 IUPAC Name: tert-butyl (E)-but-2-enoate SMILES: CC=CC(=O)OC(C)(C)C