CAS RN 79241-46-6
IUPAC Name:
butyl (2R)-2-(4-{[5-(trifluoromethyl)pyridin-2-yl]oxy}phenoxy)propanoate
Synonyms:
fluazifop-P-butyl (iso)
Molecular Weight (g/mol):
383.37
Molecular Formula:
C19H20F3NO4
InChi Key:
VAIZTNZGPYBOGF-CYBMUJFWSA-N
SMILES:
CCCCOC(=O)[C@@H](C)OC1=CC=C(OC2=CC=C(C=N2)C(F)(F)F)C=C1
1
–
1
of
1
results
1
Fluazinam, SPEX CertiPrep™
CAS: 79241-46-6 Molecular Formula: C19H20F3NO4 Molecular Weight (g/mol): 383.367 InChI Key: VAIZTNZGPYBOGF-CYBMUJFWSA-N PubChem CID: 3033674 IUPAC Name: butyl (2R)-2-[4-[5-(trifluoromethyl)pyridin-2-yl]oxyphenoxy]propanoate SMILES: CCCCOC(=O)C(C)OC1=CC=C(C=C1)OC2=NC=C(C=C2)C(F)(F)F