accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3CNNNClO

CAS RN 79307-93-0

IUPAC Name: hydrogen 4-[(4-chlorophenyl)methyl]-2-(1-methylazepan-4-yl)-1,2-dihydrophthalazin-1-one chloride
Synonyms: azelastine
Molecular Weight (g/mol): 418.36
Molecular Formula: C22H25Cl2N3O
InChi Key: YEJAJYAHJQIWNU-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CN1CCCC(CC1)N1N=C(CC2=CC=C(Cl)C=C2)C2=CC=CC=C2C1=O
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)