accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 79322-76-2

IUPAC Name: methyl 4-[(1S)-1-hydroxyethyl]benzoate
Synonyms: methyl 4-[(1S)-1-hydroxyethyl]benzoate
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: KAXLTAULVFFCNL-ZETCQYMHSA-N
SMILES: COC(=O)C1=CC=C(C=C1)[C@H](C)O
Boiling Point 
  • (2)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)