accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 79370-78-8

NHON

CAS RN 79370-78-8

IUPAC Name: 5-hydroxybenzene-1,3-dicarbonitrile
Synonyms: 5-hydroxybenzene-1,3-dicarbonitrile
Molecular Weight (g/mol): 144.13
Molecular Formula: C8H4N2O
InChi Key: CHASTCUDJNQBOZ-UHFFFAOYSA-N
SMILES: OC1=CC(=CC(=C1)C#N)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results