accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOFOH

CAS RN 79418-78-3

IUPAC Name: 3-fluoro-4-hydroxy-5-methoxybenzaldehyde
Synonyms: 3-fluoro-4-hydroxy-5-methoxybenzaldehyde
Molecular Weight (g/mol): 170.14
Molecular Formula: C8H7FO3
InChi Key: OOGOFUKAJDPHDJ-UHFFFAOYSA-N
SMILES: COC1=C(O)C(F)=CC(C=O)=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)