accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 79513-35-2

IUPAC Name: hydrogen tert-butyl N-(2-aminoethyl)carbamate chloride
Synonyms: hydrogen tert-butyl N-(2-aminoethyl)carbamate chloride
Molecular Weight (g/mol): 196.68
Molecular Formula: C7H17ClN2O2
InChi Key: XUHJJLCKXZTUJN-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC(C)(C)OC(=O)NCCN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)