accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3SBrSCH3CH3H3C

CAS RN 795274-44-1

IUPAC Name: 1-bromo-3,5-bis(tert-butylsulfanyl)benzene
Synonyms: 1-bromo-3,5-bis(tert-butylsulfanyl)benzene
Molecular Weight (g/mol): 333.35
Molecular Formula: C14H21BrS2
InChi Key: VLQVYTYMAXNJGM-UHFFFAOYSA-N
SMILES: CC(C)(C)SC1=CC(SC(C)(C)C)=CC(Br)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results