accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 79558-09-1

IUPAC Name: 2-{4-[3-(4-acetyl-3-hydroxy-2-propylphenoxy)propoxy]phenoxy}acetic acid
Synonyms: 4-[3-(4-acetyl-3-hydroxy-2-propylphenoxy)propoxy]phenoxyacetic acid
Molecular Weight (g/mol): 402.44
Molecular Formula: C22H26O7
InChi Key: HBBVCKCCQCQCTJ-UHFFFAOYSA-N
SMILES: CCCC1=C(OCCCOC2=CC=C(OCC(O)=O)C=C2)C=CC(C(C)=O)=C1O
Product Line 
  • (2)
Quantity 
  • (1)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results