accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNOOH

CAS RN 79583-98-5

IUPAC Name: 5-azidopentanoic acid
Synonyms: 5-azidopentanoic acid
Molecular Weight (g/mol): 143.15
Molecular Formula: C5H9N3O2
InChi Key: SBZDIRMBQJDCLB-UHFFFAOYSA-N
SMILES: OC(=O)CCCCN=[N+]=[N-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results