accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH2H3COHCH3H2C

CAS RN 79604-66-3

IUPAC Name: 2,4,6-trimethylhepta-1,6-dien-4-ol
Synonyms: 2,4,6-trimethylhepta-1,6-dien-4-ol
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: METKJWMTLIYYIQ-UHFFFAOYSA-N
SMILES: CC(=C)CC(C)(O)CC(C)=C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results