accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 79606-45-4

IUPAC Name: 4-chloro-N,N-bis(propan-2-yl)benzamide
Synonyms: 4-chloro-N,N-diisopropylbenzamide
Molecular Weight (g/mol): 239.74
Molecular Formula: C13H18ClNO
InChi Key: MGPPJDHMGGQMSG-UHFFFAOYSA-N
SMILES: CC(C)N(C(C)C)C(=O)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results