accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 79606-48-7

IUPAC Name: 4-nitro-N,N-bis(propan-2-yl)benzamide
Synonyms: N,N-diisopropyl-4-nitrobenzamide
Molecular Weight (g/mol): 250.3
Molecular Formula: C13H18N2O3
InChi Key: RDBKRCFDYJOIDU-UHFFFAOYSA-N
SMILES: CC(C)N(C(C)C)C(=O)C1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results