accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 79651-35-7

NNNH2Cl

CAS RN 79651-35-7

IUPAC Name: 2-(chloromethyl)pyrimidin-4-amine
Synonyms: 2-(chloromethyl)pyrimidin-4-amine
Molecular Weight (g/mol): 143.57
Molecular Formula: C5H6ClN3
InChi Key: JWAQENUKTAOJST-UHFFFAOYSA-N
SMILES: NC1=NC(CCl)=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results