accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 79669-50-4

IUPAC Name: methyl 5-bromo-2-methylbenzoate
Synonyms: methyl 5-bromo-2-methylbenzoate
Molecular Weight (g/mol): 229.07
Molecular Formula: C9H9BrO2
InChi Key: FDCYLMYCHALQJR-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Br)=CC=C1C
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)