accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 797-70-6

IUPAC Name: 1-[bis(4-methylphenyl)phosphoroso]-4-methylbenzene
Synonyms: 1-[bis(4-methylphenyl)phosphoroso]-4-methylbenzene
Molecular Weight (g/mol): 320.37
Molecular Formula: C21H21OP
InChi Key: SPKBYIYIZQARNX-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)P(=O)(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results