accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrHNN

CAS RN 79762-54-2

IUPAC Name: 6-bromo-1H-indazole
Synonyms: 6-bromo-1H-indazole
Molecular Weight (g/mol): 197.04
Molecular Formula: C7H5BrN2
InChi Key: WMKDUJVLNZANRN-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C=NNC2=C1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results