accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOCH3OCH3O

CAS RN 798-61-8

IUPAC Name: 6,7-dimethoxy-3-(4-methoxyphenyl)-4H-chromen-4-one
Synonyms: 6,7-dimethoxy-3-(4-methoxyphenyl)chromen-4-one
Molecular Weight (g/mol): 312.32
Molecular Formula: C18H16O5
InChi Key: YHXIOAVHEXKZCQ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=COC2=CC(OC)=C(OC)C=C2C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results