accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 79887-15-3

IUPAC Name: 1-butoxy-4-ethynylbenzene
Synonyms: 1-butoxy-4-ethynylbenzene
Molecular Weight (g/mol): 174.24
Molecular Formula: C12H14O
InChi Key: JMLXBCSKFMMFGF-UHFFFAOYSA-N
SMILES: CCCCOC1=CC=C(C=C1)C#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results