accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 79917-89-8

IUPAC Name: 1-methyl-3-propyl-2,3-dihydro-1H-imidazol-1-ium chloride
Synonyms: 1-methyl-3-propyl-1,2-dihydroimidazol-1-ium chloride
Molecular Weight (g/mol): 162.66
Molecular Formula: C7H15ClN2
InChi Key: GYTJXQRCNBRFGU-UHFFFAOYNA-N
SMILES: [Cl-].CCCN1C[NH+](C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results