accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFOSOOFFFFFFFFF

CAS RN 79963-95-4

IUPAC Name: 2,2,2-trifluoroethyl 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonate
Synonyms: 2,2,2-trifluoroethyl 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonate
Molecular Weight (g/mol): 382.12
Molecular Formula: C6H2F12O3S
InChi Key: KJGYBFLEIPDFNQ-UHFFFAOYSA-N
SMILES: FC(F)(F)COS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)