accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 79992-76-0

IUPAC Name: magnesium(2+) bis(2-ethylbutanoate)
Synonyms: magnesium(2+) bis(2-ethylbutanoate)
Molecular Weight (g/mol): 254.61
Molecular Formula: C12H22MgO4
InChi Key: JOADGALWHMAAKM-UHFFFAOYSA-L
SMILES: [Mg++].CCC(CC)C([O-])=O.CCC(CC)C([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results