accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 80-04-6

IUPAC Name: 4-[2-(4-hydroxycyclohexyl)propan-2-yl]cyclohexan-1-ol
Synonyms: 4-[2-(4-hydroxycyclohexyl)propan-2-yl]cyclohexan-1-ol
Molecular Weight (g/mol): 240.39
Molecular Formula: C15H28O2
InChi Key: CDBAMNGURPMUTG-UHFFFAOYSA-N
SMILES: CC(C)(C1CCC(O)CC1)C1CCC(O)CC1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)