accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OHO

CAS RN 80-15-9

IUPAC Name: 2-phenylpropane-2-peroxol
Synonyms: cumene hydroperoxide
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: YQHLDYVWEZKEOX-UHFFFAOYSA-N
SMILES: CC(C)(OO)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (8)
  • (4)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (4)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (6)
Grade 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (4)
  • (1)
  • (1)
  • (6)
  • (8)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (6)
  • (1)
  • (2)
  • (2)