accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 80-26-2

IUPAC Name: 2-(4-methylcyclohex-3-en-1-yl)propan-2-yl acetate
Synonyms: α-terpinyl acetate α-terpineol, acetate acetate α-terpineol terpinyl acetate (natural) terpineol acetate
Molecular Weight (g/mol): 196.29
Molecular Formula: C12H20O2
InChi Key: IGODOXYLBBXFDW-UHFFFAOYNA-N
SMILES: CC(=O)OC(C)(C)C1CCC(C)=CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results