accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOONH

CAS RN 80-30-8

IUPAC Name: N-cyclohexyl-4-methylbenzene-1-sulfonamide
Synonyms: N-cyclohexyl-4-methylbenzenesulfonamide
Molecular Weight (g/mol): 253.36
Molecular Formula: C13H19NO2S
InChi Key: DKYVVNLWACXMDW-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)NC1CCCCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results